Order Entry
Export
ContactUsLinkComponent
2,6-Dichlorobenzoic acid, Sigma-Aldrich®
Irritant
2,6-Dichlorobenzoic acid, Sigma-Aldrich®
Catalog # SIALD57450-25G
Supplier:  Merck
CAS Number:  
Irritant
2,6-Dichlorobenzoic acid, Sigma-Aldrich®
Catalog # SIALD57450-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :191.01 g/mol
Boiling Pt:299 °C (1013 hPa)
Melting Pt:138…139 °C
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-025-9
 :GHS07
CAS Number:50-30-6
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002418