
2,4-Dichlorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:50-84-0
:
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002414
:191.01 g/mol
Boiling Pt:310 °C (1013 hPa)
Melting Pt:158…160 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-067-8
:GHS07