
2,3-Dichlorobenzoic acid, Sigma-Aldrich®
Catalog # SIAL225339-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
Melting Pt:168.3 °C
:M-Clarity™ quality level = MQ100
:191.01 g/mol
EINECS:200-039-5
:GHS07
Storage Temperature:Ambient
CAS Number:50-45-3
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002413