3,5-Dichlorobenzhydrazide ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 62899-78-9 Molecular Formula: C7H6Cl2N2O Formula Weight: 205.04 Melting Point: 205-207° MDL No.: MFCD00833411
Synonyms: 3,5-Dichlorobenzoic hydrazide
- CAS Number:62899-78-9
- Formula:C₇H₆Cl₂N₂O
- MDL Number:MFCD00833411
- Molecular Weight:205.04 g/mol
- Melting Pt:205…207 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 62899-78-9 Molecular Formula: C7H6Cl2N2O Formula Weight: 205.04 Melting Point: 205-207° MDL No.: MFCD00833411
Synonyms: 3,5-Dichlorobenzoic hydrazide
- CAS Number:62899-78-9
- Formula:C₇H₆Cl₂N₂O
- MDL Number:MFCD00833411
- Molecular Weight:205.04 g/mol
- Melting Pt:205…207 °C
- Storage Temperature:Ambient
Documents
Organic Building Blocks and Intermediates



