2,6-Dichlorobenzamide
Catalog # EHERC12355000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:190.03 g/mol
Boiling Pt:335 °C (1013 hPa)
Boiling Pt:335 °C (1013 hPa)
Melting Pt:203 °C
Density:1,44 g/cm³ (20 °C)
EINECS:217-918-4
CAS Number:2008-58-4
Formula:C₇H₅Cl₂NO
MDL Number:MFCD00007975