2,6-Dichlorobenzamide
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:2008-58-4
:
Formula:C₇H₅Cl₂NO
MDL Number:MFCD00007975
:190.03 g/mol
Boiling Pt:335 °C (1013 hPa)
Melting Pt:203 °C
Density:1,44 g/cm³ (20 °C)
EINECS:217-918-4