Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dichlorobenzamide
2,5-Dichlorobenzamide
2,5-Dichlorobenzamide
2,5-Dichlorobenzamide
Supplier:
Thermo Scientific
Certificates
Expand image
2,5-Dichlorobenzamide
Supplier:
Thermo Scientific
About this item
CAS Number:
5980-26-7
:
Formula:
C₇H₅Cl₂NO
MDL Number:
MFCD00017609
:
190.03 g/mol