

4-Chlorobenzoyl chloride 99+%
:122-01-0
:
:C₇H₄Cl₂O
:MFCD00000686
:175.01 g/mol
:222 °C (760 mmHg)
:12…14 °C
:1.37 g/cm³
:105 °C
:Ambient
:204-515-3
:3265
:8,II
:GHS05
:GHS07
- Appearance:Clear colorless to slightly colored Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5770 to 1.5790 (20°C, 589 nm)
- GC:≥99.0 %
- Titration RCOCl:≥98.50 %