
3,4-Dichlorobenzoyl chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3024-72-4
:
Formula:C₇H₃Cl₃O
MDL Number:MFCD00000672
:209.46 g/mol
Boiling Pt:242 °C (1013 hPa)
Melting Pt:30…33 °C
Flash Pt:142 °C
Storage Temperature:Ambient
EINECS:221-177-2
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to yellow Low melting crystalline solid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Titration RCOCl:≥96.0 %