

2,6-Dichlorobenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4659-45-4
:
Formula:C₇H₃Cl₃O
MDL Number:MFCD00000662
:209.46 g/mol
Boiling Pt:260 °C (1013 hPa)
Melting Pt:18 °C
Density:1.45 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:225-102-4
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:3265
:GHS05
:GHS07