

2,6-Dichlorobenzoyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4659-45-4
:
Formula:C₇H₃Cl₃O
MDL Number:MFCD00000662
:209.46 g/mol
Boiling Pt:130…132 °C (15 mmHg)
Melting Pt:15…18 °C
Density:1.463
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:225-102-4
UN:3265
ADR:8,II
:GHS05
:GHS07