

2,4-Dichlorobenzoyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89-75-8
:
Formula:C₇H₃Cl₃O
MDL Number:MFCD00000661
:209.46 g/mol
Boiling Pt:150 °C (34 mmHg)
Melting Pt:16…20 °C
Density:1.497
Flash Pt:137 °C (278 °F)
Storage Temperature:Ambient
EINECS:201-936-4
UN:3265
ADR:8,II
:GHS05
:GHS07