

2,4-Dichlorobenzoyl chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89-75-8
:
Formula:C₇H₃Cl₃O
MDL Number:MFCD00000661
:209.46 g/mol
Boiling Pt:150 °C (34 mmHg)
Melting Pt:21 °C
Density:1.49 g/cm³
Flash Pt:137 °C
Storage Temperature:Ambient
EINECS:201-936-4
UN:3261
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Clear colorless to slightly colored Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.5915 to 1.5935 (20°C, 589 nm)
- GC:≥97.5 %
- Titration RCOCl:≥97.5 %