3,4-Dichlorobenzonitrile, Sigma-Aldrich®
SIAL532177-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:172.01 g/mol
:86…89 °C
:Ambient
:3439
:6.1,III
:M-Clarity™ quality level = MQ100
:229-494-8
:6574-99-8
:C₇H₃Cl₂N
:MFCD00016379