Order Entry
Export
ContactUsLinkComponent
4-Chloro-3-nitrobenzaldehyde, Sigma-Aldrich®
4-Chloro-3-nitrobenzaldehyde, Sigma-Aldrich®
Catalog # SIALC58703-25G
Supplier:  Merck
CAS Number:  
4-Chloro-3-nitrobenzaldehyde, Sigma-Aldrich®
Catalog # SIALC58703-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :185.57 g/mol
Melting Pt:57…60 °C
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:240-645-7
CAS Number:16588-34-4
Formula:C₇H₄ClNO₃
MDL Number:MFCD00007078