Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dichlorobenzaldehyde
2,5-Dichlorobenzaldehyde, Sigma-Aldrich®
2,5-Dichlorobenzaldehyde, Sigma-Aldrich®
2,5-Dichlorobenzaldehyde, Sigma-Aldrich®
SIAL523917-5G
:
Merck
:
2,5-Dichlorobenzaldehyde, Sigma-Aldrich®
SIAL523917-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
M-Clarity™ quality level = MQ100
:
175.01 g/mol
:
228-832-1
:
GHS07
:
6361-23-5
:
C₇H₄Cl₂O
:
MFCD00156140