Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichlorophenylboronic acid
3,5-Dichlorophenylboronic acid ≥98%
3,5-Dichlorophenylboronic acid ≥98%
Irritant
3,5-Dichlorophenylboronic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
3,5-Dichlorophenylboronic acid ≥98%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
67492-50-6
:
Formula:
C₆H₅BCl₂O₂
MDL Number:
MFCD00051935
:
190.82 g/mol
Melting Pt:
∼ 315 °C (dec.)
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAB22765
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation