Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dichlorophenylboronic acid
3,4-Dichlorophenylboronic acid, Sigma-Aldrich®
3,4-Dichlorophenylboronic acid, Sigma-Aldrich®
Irritant
3,4-Dichlorophenylboronic acid, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Irritant
3,4-Dichlorophenylboronic acid, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
151169-75-4
:
Formula:
C₆H₅BCl₂O₂
MDL Number:
MFCD01074646
:
190.82 g/mol
:
M-Clarity™ quality level = MQ100
:
GHS07