Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Chlorophenylboronic acid 97%

Irritant
2-Chlorophenylboronic acid 97%
Certificates
2-Chlorophenylboronic acid 97%

2 Options Available Below

Product Details & Documents
Synonyms:  (2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid
  • CAS Number:3900-89-8
  • Formula:C₆H₆BClO₂
  • MDL Number:MFCD00674012
  • Molecular Weight:156.38 g/mol
  • Melting Pt:94…95 °C
  • Storage Temperature:Refrigerator

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light beige Powder or crystals or granules
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 42,30 €
    42,30 €
    1 * 1 g
       42,30 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 138,00 €
    138,00 €
    1 * 5 g
       138,00 €