Order Entry
Export
Orders LinkContactUsLinkComponent
 

Hexachlorobenzene

Glass bottle for solids, 1 g
Catalog # MAYBBTB15040.1
Health
Environment
Supplier:   Thermo Scientific
Hexachlorobenzene
Catalog # MAYBBTB15040.1
Supplier:  Thermo Scientific
CAS Number:  118-74-1
Hexachlorobenzene

Glass bottle for solids, 1 g

CAS Number:  118-74-1
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:118-74-1
  • Formula:C₆Cl₆
  • Molecular Weight:284.78 g/mol
  • Merck Index:12,04714
  • Boiling Pt:332 °C (1013 hPa)
  • Melting Pt:2.045 °C
  • Density:2.045 g/cm³ (20 °C)
  • Flash Pt:242 °C
  • Storage Temperature:Ambient
  • EINECS:204-273-9
  • ADR:6.1,III
  •  :Density: 2.045 g/cm³ (20 °C)
  • UN:2729
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications