1,3-Dichlorobenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:541-73-1
:
Formula:C₆H₄Cl₂
MDL Number:MFCD00000573
Merck Index:13,03080
Boiling Pt:173 °C (1013 hPa)
Melting Pt:–25 °C
Density:1.2884 g/cm³ (20 °C)
Flash Pt:65 °C
EINECS:208-792-1
UN:3082
ADR:9,III
:M-Clarity™ quality level = MQ100
:147 g/mol
:GHS07
:GHS09
Storage Temperature:Ambient