Order Entry
Export
Orders LinkContactUsLinkComponent
3,4-Dihydroxybenzaldehyde

3,4-Dihydroxybenzaldehyde

Glass bottle for solids, 25 g
Catalog # MOLEM31371877
Irritant
Supplier:   Molekula
3,4-Dihydroxybenzaldehyde
Catalog # MOLEM31371877
Supplier:  Molekula
CAS Number:  139-85-5
3,4-Dihydroxybenzaldehyde

Glass bottle for solids, 25 g

CAS Number:  139-85-5
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Protocatechuic aldehyde, Protocatechualdehyde
  • CAS Number:139-85-5
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00003370
  • Molecular Weight:138.12 g/mol
  • Merck Index:12,08079
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:150…155 °C
  • Density:1.505 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-377-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications