Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dihydroxybenzaldehyde

2,3-Dihydroxybenzaldehyde

Glass bottle for solids, 1 g
Catalog # ROTH7274.1
Irritant
Supplier:   Roth Carl
2,3-Dihydroxybenzaldehyde
Catalog # ROTH7274.1
Supplier:  Roth Carl
CAS Number:  24677-78-9
2,3-Dihydroxybenzaldehyde

Glass bottle for solids, 1 g

CAS Number:  24677-78-9
Price
20,80 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:24677-78-9
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00003324
  • Molecular Weight:138.12 g/mol
  • Boiling Pt:235 °C (1013 hPa)
  • Melting Pt:103…105 °C
  • Storage Temperature:Ambient
  • EINECS:246-398-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications