2,4,6-Trimethoxybenzaldehyde, Sigma-Aldrich®
SIAL138711-10G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
:
:196.2 g/mol
:291 °C (1013 hPa)
:120 °C
:M-Clarity™ quality level = MQ100
:Ambient
:212-598-2
:830-79-5
:C₁₀H₁₂O₄
:MFCD00003313