2,4-Dinitrobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
- CAS Number:528-75-6
- Formula:C₇H₄N₂O₅
- MDL Number:MFCD00013376
- Molecular Weight:196.12 g/mol
- Merck Index:13,03300
- Boiling Pt:378 °C (1013 hPa)
- Melting Pt:72 °C
- :M-Clarity™ quality level = MQ200
- EINECS:208-440-7
3 Options Available Below
Product Details & Documents
- CAS Number:528-75-6
- Formula:C₇H₄N₂O₅
- MDL Number:MFCD00013376
- Molecular Weight:196.12 g/mol
- Merck Index:13,03300
- Boiling Pt:378 °C (1013 hPa)
- Melting Pt:72 °C
- :M-Clarity™ quality level = MQ200
- EINECS:208-440-7



