Order Entry
Export
ContactUsLinkComponent
4-Cyanobenzaldehyde
4-Cyanobenzaldehyde
Catalog # MOLE24240192-25G
Supplier:  Molekula
CAS Number:  
4-Cyanobenzaldehyde
Catalog # MOLE24240192-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :131.13 g/mol
Boiling Pt:262 °C (1013 hPa)
Melting Pt:100…103 °C
Storage Temperature:Ambient
EINECS:203-267-3
UN:3439
ADR:6.1,III
CAS Number:105-07-7
Formula:C₈H₅NO
MDL Number:MFCD00003376