Order Entry
Export
ContactUsLinkComponent
4-Cyanobenzaldehyde 98%
4-Cyanobenzaldehyde 98%
Certificates
undefined
4-Cyanobenzaldehyde 98%

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    98 to 103 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information