2,4-Dibromophenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:615-58-7
:
Formula:C₆H₄Br₂O
MDL Number:MFCD00002149
:251.91 g/mol
Boiling Pt:238…239 °C (1013 hPa)
Melting Pt:38 °C
Density:2.07 g/cm³ (20 °C)
EINECS:210-436-5
ADR:6.1,II
:M-Clarity™ quality level = MQ100
UN:2811