2,5-Dibromoaniline, Sigma-Aldrich®
Catalog # SIAL125881-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:250.92 g/mol
Boiling Pt:281 °C (1013 hPa)
Melting Pt:54.5…55 °C
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:222-865-5
UN:2811
CAS Number:3638-73-1
Formula:C₆H₅Br₂N
MDL Number:MFCD00007636