
2-Bromo-4,5-difluorophenol, Sigma-Aldrich®
Catalog # SIAL369071-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:208.99 g/mol
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:166281-37-4
Formula:C₆H₃BrF₂O
MDL Number:MFCD00070751