Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2,3-difluorophenol
5-Bromo-2,3-difluorophenol ≥97%
5-Bromo-2,3-difluorophenol ≥97%
:
Thermo Fisher Scientific
5-Bromo-2,3-difluorophenol ≥97%
:
Thermo Fisher Scientific
:
186590-26-1
:
:
C₆H₃BrF₂O
:
MFCD01631350
:
208.99 g/mol
:
GHS06
:
GHS07
:
Ambient
msds_ALFAB23853
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years