Order Entry
Export
ContactUsLinkComponent
4-Bromo-2,3-difluorophenol ≥97%
Toxic
Irritant
4-Bromo-2,3-difluorophenol ≥97%
Catalog # H26369.06
CAS Number:  
Toxic
Irritant
4-Bromo-2,3-difluorophenol ≥97%
Catalog # H26369.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:144292-32-0
 :
Formula:C₆H₃BrF₂O
MDL Number:MFCD08061907
 :208.99 g/mol
EINECS:None
 :GHS06
 :GHS07
Storage Temperature:Ambient