

2-Bromo-4,6-difluoroaniline, Sigma-Aldrich®
Catalog # SIAL297690-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:208.01 g/mol
Boiling Pt:227 °C (1013 hPa)
Melting Pt:42…42.5 °C
ADR:4.1,II
:M-Clarity™ quality level = MQ100
EINECS:429-430-0
UN:1325
:GHS07
:GHS09
CAS Number:444-14-4
Formula:C₆H₄BrF₂N
MDL Number:MFCD00009639