

2-Bromo-4,6-difluoroaniline
Supplier: CHEMPUR
Certificates
About this item
CAS Number:444-14-4
:
Formula:C₆H₄BrF₂N
MDL Number:MFCD00009639
:208.01 g/mol
Boiling Pt:227 °C (1013 hPa)
Melting Pt:42…42.5 °C
EINECS:429-430-0
UN:1325
ADR:4.1,II
:GHS07
:GHS09