

1-Bromo-2,6-difluorobenzene, Sigma-Aldrich®
Catalog # SIAL263451-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:192.99 g/mol
Boiling Pt:61 °C (1013 hPa)
Density:1.71 g/cm³ (20 °C)
ADR:3,III
:M-Clarity™ quality level = MQ100
EINECS:264-750-2
UN:1993
:GHS02
:GHS07
CAS Number:64248-56-2
Formula:C₆H₃BrF₂
MDL Number:MFCD00009894