Order Entry
Export
ContactUsLinkComponent
1,2-Dibromotetrafluorobenzene, Sigma-Aldrich®
Irritant
1,2-Dibromotetrafluorobenzene, Sigma-Aldrich®
Catalog # SIAL100161-5G
Supplier:  Merck
CAS Number:  
Irritant
1,2-Dibromotetrafluorobenzene, Sigma-Aldrich®
Catalog # SIAL100161-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :307.87 g/mol
Boiling Pt:93…96 °C (25 mmHg)
Melting Pt:12 °C
Density:2.23 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:212-564-7
 :GHS07
CAS Number:827-08-7
Formula:C₆Br₂F₄
MDL Number:MFCD00039209