

1,2-Dibromo-4,5-difluorobenzene, Sigma-Aldrich®
Catalog # SIAL264237-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
Boiling Pt:208 °C (1013 hPa)
Melting Pt:32.5…33.5 °C
EINECS:265-021-1
ADR:4.1,II
:M-Clarity™ quality level = MQ100
:271.89 g/mol
UN:1325
:GHS02
:GHS07
CAS Number:64695-78-9
Formula:C₆H₂Br₂F₂
MDL Number:MFCD00009890