
3,5-Dibromosalicylaldehyde, Sigma-Aldrich®
Catalog # SIAL122130-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:279.92 g/mol
Merck Index:13,03052
Boiling Pt:261 °C (1013 hPa)
Melting Pt:86 °C
:M-Clarity™ quality level = MQ200
EINECS:202-003-4
:GHS07
CAS Number:90-59-5
Formula:C₇H₄Br₂O₂
MDL Number:MFCD00003318