5-Bromovanillin, Sigma-Aldrich®
Catalog # SIAL130605-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:231.05 g/mol
Boiling Pt:314 °C (1013 hPa)
Melting Pt:166…167 °C
:M-Clarity™ quality level = MQ100
EINECS:221-016-6
:GHS07
CAS Number:2973-76-4
Formula:C₈H₇BrO₃
MDL Number:MFCD00006940