
3,4-Dimethoxyphenylboronic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122775-35-3
:
Formula:C₈H₁₁BO₄
MDL Number:MFCD01074574
:181.98 g/mol
Melting Pt:238…240 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to beige Powder or granules and/or chunks
- Infrared spectrum:Conforms
- HPLC:≥96.0 %