2,5-Dimethoxyphenylboronic acid
Catalog # MAYBAC35921.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.98 g/mol
CAS Number:107099-99-0
Formula:C₈H₁₁BO₄
MDL Number:MFCD01318181