1,3-Benzodioxole, Sigma-Aldrich®
SIAL159166-50G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
:
:122.12 g/mol
:172 °C (1013 hPa)
:–18 °C
:1.18 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:205-992-0
:GHS07
:274-09-9
:C₇H₆O₂
:MFCD00005818