Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Benzodioxole

1,3-Benzodioxole

Irritant
Supplier:   Thermo Scientific
1,3-Benzodioxole
Supplier:  Thermo Scientific
Certificates
1,3-Benzodioxole

3 Options Available Below

Product Details & Documents
Synonyms:  Benzo[d][1,3]dioxole, 1,2-Methylenedioxybenzene
  • CAS Number:274-09-9
  • Formula:C₇H₆O₂
  • MDL Number:MFCD00005818
  • Molecular Weight:122.12 g/mol
  • Boiling Pt:172 °C (1013 hPa)
  • Melting Pt:–18 °C
  • Density:1,18 g/cm³ (20 °C)
  • EINECS:205-992-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g