1,3-Benzodioxole ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 274-09-9 Molecular Formula: C7H6O2 Formula Weight: 122.12 Flash Point: 55°(131°F) Refractive Index: 1.5390 Odour: Aromatic Melting Point: -18° Boiling Point: 172-173° Beilstein Reference: 115506 MDL No.: MFCD00005818
Synonyms: Benzo[d][1,3]dioxole, 1,2-Methylenedioxybenzene
- CAS Number:274-09-9
- Formula:C₇H₆O₂
- MDL Number:MFCD00005818
- Molecular Weight:122.12 g/mol
- Boiling Pt:172…173 °C
- Melting Pt:–18 °C
- Density:1.185
- Flash Pt:55 °C (131 °F)
- Storage Temperature:Ambient
- EINECS:205-992-0
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 274-09-9 Molecular Formula: C7H6O2 Formula Weight: 122.12 Flash Point: 55°(131°F) Refractive Index: 1.5390 Odour: Aromatic Melting Point: -18° Boiling Point: 172-173° Beilstein Reference: 115506 MDL No.: MFCD00005818
Synonyms: Benzo[d][1,3]dioxole, 1,2-Methylenedioxybenzene
- CAS Number:274-09-9
- Formula:C₇H₆O₂
- MDL Number:MFCD00005818
- Molecular Weight:122.12 g/mol
- Boiling Pt:172…173 °C
- Melting Pt:–18 °C
- Density:1.185
- Flash Pt:55 °C (131 °F)
- Storage Temperature:Ambient
- EINECS:205-992-0



