
1,2-Naphthoquinone 95% tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:524-42-5
:
Formula:C₁₀H₆O₂
MDL Number:MFCD00001698
:158.16 g/mol
Merck Index:12,06481
Boiling Pt:296 °C (1013 hPa)
Melting Pt:136…141 °C
Density:1,45 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-360-2
ADR:6.1,II
:GHS07
Specification Test Results
- Appearance:Brown Powder
- Infrared spectrum:Conforms
- GC:≥94 %