

1,4-Naphthoquinone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:130-15-4
:
Formula:C₁₀H₆O₂
MDL Number:MFCD00001676
:158.16 g/mol
Merck Index:12,06482
Boiling Pt:298 °C (1013 hPa)
Melting Pt:124 °C
Density:1.422 g/cm³ (20 °C)
EINECS:204-977-6
UN:2811
ADR:6.1,I
:M-Clarity™ quality level = MQ200
Flash Pt:141 °C
:GHS06
:GHS09