


1,4-Naphthoquinone, (max. 6% H₂O) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:130-15-4
:
Formula:C₁₀H₆O₂
MDL Number:MFCD00001676
:158.16 g/mol
Merck Index:12,06482
Boiling Pt:298 °C (1013 hPa)
Melting Pt:128.5 °C
Density:1,422 g/cm³ (20 °C)
Flash Pt:141 °C
Storage Temperature:Ambient
EINECS:204-977-6
UN:2928
ADR:6.1,I
:GHS05
:GHS06
:GHS09
Specification Test Results
- Appearance:Khaki Powder
- Infrared spectrum:Conforms
- Water:≤6 % (K.F.)
- GC:≥99 %
- HPLC:≥97.5 %