
7-Isopropyl-1,4-dimethylazulene
Supplier: Roth Carl
Certificates
About this item
CAS Number:489-84-9
:
Formula:C₁₅H₁₈
MDL Number:MFCD00003811
:198.31 g/mol
Merck Index:12,04581
Boiling Pt:309 °C (1013 hPa)
Melting Pt:133 °C
Density:0.9759 g/cm³ (18 °C)
EINECS:207-701-2
:GHS07