Piperidine
Glass bottle, 200 mL
Catalog # ROTHA122.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Hexahydropyridine, Hexazane, Pentamethyleneimine
- CAS Number:110-89-4
- Formula:C₅H₁₁N
- MDL Number:MFCD00005979
- Molecular Weight:85.15 g/mol
- Merck Index:12,07621
- Boiling Pt:107.3 °C (1013 hPa)
- Melting Pt:–10 °C
- Density:0.8622 g/cm³ (20 °C)
- Flash Pt:16 °C
- Storage Temperature:Ambient
- EINECS:203-813-0
- UN:2401
- ADR:8,I
Specifications
Specifications
- Pack type:Glass bottle
- Pk:200 mL



