3,3-Dimethyl-1-butyne, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:917-92-0
:
Formula:C₆H₁₀
MDL Number:MFCD00008852
:82.15 g/mol
Boiling Pt:38 °C (1013 hPa)
Melting Pt:–78.2 °C
Density:1.0003 g/cm³ (0 °C)
Storage Temperature:Refrigerator
EINECS:213-035-3
ADR:3,II
:M-Clarity™ quality level = MQ200
UN:3295